Learn More
Aviva Systems Biology Corp Foxd4 Antibody - N-terminal region (ARP36855_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36855_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xd4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of mouse Foxd4. Alternate names: FREAC5, Fkh2ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-526