Learn More
Aviva Systems Biology Corp FOXE3 Antibody - C-terminal region (ARP32304_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32304_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XE3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FOXE3. Alternate names: FKHL12, FREAC8ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-54521