Learn More
Aviva Systems Biology Corp FOXF1 Antibody - N-terminal region (ARP31691_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31691_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXF1. Alternate names: FKHL5, ACDMPV, FREAC1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-54306