Learn More
Aviva Systems Biology Corp FOXF1A Antibody - C-terminal region (ARP37047_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37047_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XF1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Hfh8, Foxf1, HFH-8, FREAC1, Freac-1, AI450827, FOXF1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56316