Learn More
Aviva Systems Biology Corp Foxf1a Antibody - middle region (ARP37048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xf1a. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Foxf1a. Alternate names: AI450827, FREAC1, Foxf1, Freac-1, HFH-8, Hfh8, Foxf1aApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-573