Learn More
Aviva Systems Biology Corp FOXH1 Antibody - middle region (ARP36853_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36853_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse FOXH1. Alternate names: Fast2ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56285