Learn More
Aviva Systems Biology Corp FOXI1 Antibody - C-terminal region (ARP38964_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38964_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FOXI1. Alternate names: FKHL10, FREAC6, HFH3, MGC34197, FKH10, HFH-3, FREAC-6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56869