Learn More
Aviva Systems Biology Corp Foxi2 Antibody - N-terminal region (ARP37604_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37604_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xi2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: B130055A05RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-667