Learn More
Aviva Systems Biology Corp FOXJ1 Antibody - middle region (ARP35642_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XJ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOXJ1. Alternate names: FKHL13, HFH-4, HFH4, MGC35202ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55799