Learn More
Aviva Systems Biology Corp Foxk1 Antibody - middle region (ARP37942_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37942_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xk1. It was validated on Western Blot. .xk1,forkhead box K1,Mnf; Gm10868; AI463295; A630048H08Rik; ENSMUSG00000075577;.xk1,forkhead box protein K1,forkhead box protein K1,myocyte nuclear factorThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: A630048H08Rik, AI463295, Mnf, Gm10868, ENSMUSG00000075577ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-704