Learn More
ABclonal Technology FOXK1 Rabbit pAb
SDP
Catalog No. 5000108651
Encompass_Preferred
Enables 14-3-3 protein binding activity, DNA-binding transcription repressor activity, RNA polymerase II-specific, and transcription cis-regulatory region binding activity. Involved in several processes, including cellular glucose homeostasis, negative regulation of autophagy, and regulation of transcription, DNA-templated. Located in cytoplasm and nucleus.
Catalog No. 50-001-08651 Supplier ABclonal Technology Supplier No. A152201000UL
Enables 14-3-3 protein binding activity, DNA-binding transcription repressor activity, RNA polymerase II-specific, and transcription cis-regulatory region binding activity. Involved in several processes, including cellular glucose homeostasis, negative regulation of autophagy, and regulation of transcription, DNA-templated. Located in cytoplasm and nucleus.