Learn More
Aviva Systems Biology Corp FOXK2 Antibody - C-terminal region (P100774_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100774_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FOXK2. Alternate names: ILF, ILF1, ILF-1ApplicationWB. Species reactivity: Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62707