Learn More
Aviva Systems Biology Corp FOXK2 Antibody - C-terminal region (ARP40032_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40032_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FOXK2. Alternate names: ILF, ILF1, ILF-1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57272