Learn More
Aviva Systems Biology Corp FOXL2 Antibody - N-terminal region (ARP33357_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33357_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXL2. Alternate names: BPES, PFRK, POF3, BPES1, PINTOApplicationWB. Species reactivity: Cow, Dog, Goat, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54978