Learn More
Aviva Systems Biology Corp Foxl2 Antibody - C-terminal region (ARP39574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xl2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Foxl2ApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-816