Learn More
Aviva Systems Biology Corp FOXM1 Antibody - N-terminal region (ARP39518_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39518_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXM1. Alternate names: FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TGT3, TRIDENTApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57069