Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology FoxM1 Monoclonal Antibody, Rabbit, Human, 20 μL
SDP

Catalog No. 502733115
Encompass

This antibody detects endogenous levels of total FoxM1 protein and is predicted to recognize all three isoforms, including FoxM1a, FoxM1b and FoxM1c. The monoclonal antibody is produced by immunizing animals with recombinant protein to human FoxM1b.

  • Reactivity: Human
  • Applications: Western blotting and immunofluorescence (immunocytochemistry)
  • Molecular weight: 110 kDa
  • Host species: Rabbit
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 µg/ml BSA, 50% glycerol and less than 0.02% sodium azide

Catalog No. 50-273-3115 Supplier Cell Signaling Technology Supplier No. 5436T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody detects endogenous levels of total FoxM1 protein and is predicted to recognize all three isoforms, including FoxM1a, FoxM1b and FoxM1c. The monoclonal antibody is produced by immunizing animals with recombinant protein to human FoxM1b.

  • Reactivity: Human
  • Applications: Western blotting and immunofluorescence (immunocytochemistry)
  • Molecular weight: 110 kDa
  • Host species: Rabbit
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 µg/ml BSA, 50% glycerol and less than 0.02% sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.