Learn More
Aviva Systems Biology Corp Foxn2 Antibody - N-terminal region (ARP37593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xn2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 3230402J05Rik, 6030465J18Rik, Fkh19, Foxn1, HTLFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-664