Learn More
Aviva Systems Biology Corp FOXN3 Antibody - middle region (ARP32840_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32840_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOXN3. Alternate names: C14orf116, CHES1, PRO1635ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54789