Learn More
ABclonal Technology FOXN3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15039500UL
Encompass_Preferred
This gene is a member of the forkhead/winged helix transcription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Checkpoint suppressor 1 suppresses multiple yeast checkpoint mutations including mec1, rad9, rad53 and dun1 by activating a MEC1-independent checkpoint pathway. Alternative splicing is observed at the locus, resulting in distinct isoforms.
Catalog No. 50-001-08213