Learn More
Aviva Systems Biology Corp FOXO1A Antibody - N-terminal region (ARP38094_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38094_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XO1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXO1A. Alternate names: FKH1, FKHR, FOXO1AApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-56517