Learn More
Aviva Systems Biology Corp FOXO3 Antibody - N-terminal region (ARP38040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XO3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXO3. Alternate names: AF6q21, DKFZp781A0677, FKHRL1, FKHRL1P2, FOXO2, FOXO3A, MGC12739, MGC31925ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56497