Learn More
Aviva Systems Biology Corp Foxo4 Antibody - C-terminal region (ARP38710_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38710_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xo4. It was validated on Western Blot. . Alternate names: Afxh, MGC117660, Mllt7, afxApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-754