Learn More
Aviva Systems Biology Corp FOXO4 Antibody - middle region (AVARP01018_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP01018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XO4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOXO4. Alternate names: AFX, AFX1, MGC120490, MLLT7ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62467