Learn More
Aviva Systems Biology Corp FOXO6 Antibody - N-terminal region (ARP37610_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37610_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XO6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53083