Learn More
Cell Signaling Technology FoxP3 Monoclonal Antibody, PE Conjugate, Mouse Reactivity, 100 μg
SDP
Catalog No. 502732290
Encompass
This monoclonal antibody is conjugated to PE and is validated for the direct flow cytometric analysis of endogenous FoxP3 protein in mouse cells. The antibody specifically targets an epitope within the intracellular domain of the transcription factor and is purified via affinity chromatography from tissue culture supernatant.
  • Validated for flow cytometry (fixed/permeabilized) applications.
  • Conjugated to PE under optimized conditions with unreacted dye removed.
  • Recognizes endogenous levels of total protein.
  • Supplied in 10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, at pH 7.2.
  • Recommended for use at a dilution of 1:160, with an optimal test amount of 0.125 μg.
Catalog No. 50-273-2290 Supplier Cell Signaling Technology Supplier No. 65210S
This monoclonal antibody is conjugated to PE and is validated for the direct flow cytometric analysis of endogenous FoxP3 protein in mouse cells. The antibody specifically targets an epitope within the intracellular domain of the transcription factor and is purified via affinity chromatography from tissue culture supernatant.
  • Validated for flow cytometry (fixed/permeabilized) applications.
  • Conjugated to PE under optimized conditions with unreacted dye removed.
  • Recognizes endogenous levels of total protein.
  • Supplied in 10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, at pH 7.2.
  • Recommended for use at a dilution of 1:160, with an optimal test amount of 0.125 μg.