Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Catalog No. 501885035
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: FXP3/197. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Tonsil or Testis. Cellular Staining: Nuclear, Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length FOXP3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 47-55kDa.Chromosome: Xp11.23. Gene Symbol: FOXP3. Gene ID: 50943. Ensemble: ENSG00000049768. Uniprot: NP_054728.
Catalog No. 50-188-5035 Supplier enQuire Bioreagents Supplier No. 50943MSM2P1
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: FXP3/197. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Tonsil or Testis. Cellular Staining: Nuclear, Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length FOXP3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 47-55kDa.Chromosome: Xp11.23. Gene Symbol: FOXP3. Gene ID: 50943. Ensemble: ENSG00000049768. Uniprot: NP_054728.