Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 50943MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM579. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Tonsil, lymph node or Breast carcinoma. Cellular Staining: Predominantly nuclear, some cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Full-length human FOXP3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 47-55kDa.Chromosome: Xp11.23. Gene Symbol: FOXP3. Gene ID: 50943. Ensemble: ENSG00000049768. Uniprot: NP_054728.
Catalog No. 50-188-5036