Learn More
Aviva Systems Biology Corp FOXP3 Antibody - N-terminal region (ARP32743_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32743_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XP3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FOXP3. Alternate names: JM2, AIID, IPEX, PIDX, XPID, DIETERApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54737