Learn More
Leica Biosystems FOXP3 BOND RTU PRIMARY 7ML
Catalog No. NC2746239
Encompass
Antigen BackgroundFOXP3 (Forkhead Box Protein 3, Scurfin) is a member of the forkhead box (FOX) famly of transcription factors. These proteins are characterised by a highly-conserved forkhead DNA-binding domain. FOXP3 has an important role in immune regulation, with its effect in reducing Interleukin-2 expression causing a reduced function of T-helper cells. The role of FOXP3 in cancer is multifaceted, with roles in both tumour progression (via suppression of T-cell activity) and oncogene suppression.DisclaimerFOXP3 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. NC2746239 Supplier Leica Biosystems Supplier No. PA0263U
Antigen BackgroundFOXP3 (Forkhead Box Protein 3, Scurfin) is a member of the forkhead box (FOX) famly of transcription factors. These proteins are characterised by a highly-conserved forkhead DNA-binding domain. FOXP3 has an important role in immune regulation, with its effect in reducing Interleukin-2 expression causing a reduced function of T-helper cells. The role of FOXP3 in cancer is multifaceted, with roles in both tumour progression (via suppression of T-cell activity) and oncogene suppression.DisclaimerFOXP3 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.