Learn More
Aviva Systems Biology Corp FOXP4 Antibody - N-terminal region (ARP39816_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39816_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXP4. Alternate names: FLJ40908, FLJ44184, hFKHLAApplicationWB. Species reactivity: Human
Catalog No. 50-014-57188