Learn More
Aviva Systems Biology Corp FOXQ1 Antibody - N-terminal region (ARP39755_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39755_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XQ1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FOXQ1. Alternate names: HFH1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57170