Learn More
Aviva Systems Biology Corp Foxq1 Antibody - middle region (ARP36873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xq1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Foxq1. Alternate names: HFH-1, Hfh1, Hfh1l, saApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-531