Learn More
Aviva Systems Biology Corp FOXR1 Antibody - N-terminal region (ARP47738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXR1. Alternate names: DLNB13, FOXN5, MGC149486ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53530