Learn More
ABclonal Technology FOXS1 Rabbit pAb
SDP
Catalog No. 5000107808
Encompass_Preferred
The forkhead family of transcription factors belongs to the winged helix class of DNA-binding proteins. The protein encoded by this intronless gene contains a forkhead domain and is found predominantly in aorta and kidney. The function of the encoded protein is unknown.
Catalog No. 50-001-07808 Supplier ABclonal Technology Supplier No. A14732500UL
The forkhead family of transcription factors belongs to the winged helix class of DNA-binding proteins. The protein encoded by this intronless gene contains a forkhead domain and is found predominantly in aorta and kidney. The function of the encoded protein is unknown.