Learn More
Cayman Chemical Foxy-5 (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502989011
Encompass_Preferred
A Wnt5a peptide mimetic with anticancer activity; increases adhesion and reduces cell motility of MDA-MB-468 breast cancer cells from 1-1,000 µM; reduces the number of lymph node metastases without affecting primary tumor growth in a DU145-Luc mouse xenograft model at 2 mg/kg
Catalog No. 50-298-9011 Supplier Cayman Chemical Supplier No. 309835 MG
A Wnt5a peptide mimetic with anticancer activity; increases adhesion and reduces cell motility of MDA-MB-468 breast cancer cells from 1-1,000 µM; reduces the number of lymph node metastases without affecting primary tumor growth in a DU145-Luc mouse xenograft model at 2 mg/kg