Learn More
Santa Cruz Biotechnology FP-Biotin (CAS 259270-28-5) 1mg Purity: ≥98% Molecular Weight: 592.75Molecular Formula:C27H50FN4O5PS
Catalog No. NC3254867
Encompass
FP-Biotin is a biotinylated organophosphorus agent that may be useful in identifying proteins that react with organophosphorus toxicants (OP). It is a small molecule that is composed of a biotin molecule attached to a fluorescent dye, fluorescein. FP-biotin has a wide range of applications in scientific research, as it can be used for labeling, imaging, and detection of molecules. It is also used in a variety of laboratory experiments, such as protein purification, enzyme assays, and DNA sequencing. Common techniques it is used in are western blotting, ELISA, and affinity chromatography.
Catalog No. NC3254867 Supplier Santa Cruz Biotechnology Supplier No. SC215056A
FP-Biotin is a biotinylated organophosphorus agent that may be useful in identifying proteins that react with organophosphorus toxicants (OP). It is a small molecule that is composed of a biotin molecule attached to a fluorescent dye, fluorescein. FP-biotin has a wide range of applications in scientific research, as it can be used for labeling, imaging, and detection of molecules. It is also used in a variety of laboratory experiments, such as protein purification, enzyme assays, and DNA sequencing. Common techniques it is used in are western blotting, ELISA, and affinity chromatography.