Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FP-Biotin | 259270-28-5 | 99.1% | 592.75 | 1 MG
SDP

Catalog No. 5000348490
Encompass_Preferred

FP-biotin is a potent organophosphorus toxicant, ideal for identifying new biomarkers of organophosphorus toxicants exposure. It quantifies the activity of FAAH, ABHD6, and MAG-lipase. This product is utilized in plasma studies because biotinylated peptides can be easily purified by binding to immobilized avidin beads. It is suitable for identifying organophosphorus toxicants reactive proteins and is for research use only.

  • Quantifies FAAH, ABHD6, and MAG-lipase activity
  • Used in plasma studies for easy purification of biotinylated peptides
  • Suitable for identifying organophosphorus toxicants reactive proteins
  • Appears as a white to light yellow solid
  • Ships at room temperature in continental US
  • Store at -20°C under nitrogen; in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO (25 mg/mL)

Catalog No. 50-003-48490 Supplier Medchemexpress LLC Supplier No. HY1369241MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FP-biotin is a potent organophosphorus toxicant, ideal for identifying new biomarkers of organophosphorus toxicants exposure. It quantifies the activity of FAAH, ABHD6, and MAG-lipase. This product is utilized in plasma studies because biotinylated peptides can be easily purified by binding to immobilized avidin beads. It is suitable for identifying organophosphorus toxicants reactive proteins and is for research use only.

  • Quantifies FAAH, ABHD6, and MAG-lipase activity
  • Used in plasma studies for easy purification of biotinylated peptides
  • Suitable for identifying organophosphorus toxicants reactive proteins
  • Appears as a white to light yellow solid
  • Ships at room temperature in continental US
  • Store at -20°C under nitrogen; in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO (25 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.