Learn More
Medchemexpress LLC FP-Biotin | 259270-28-5 | 99.1% | 592.75 | 10 MM * 1 ML
SDP
Catalog No. 5000350282
Encompass_Preferred
FP-Biotin is a potent organophosphorus toxicant suitable for identifying new biomarkers of organophosphorus toxicant exposure. It quantifies the activity of FAAH, ABHD6, and MAG-lipase. It is used in plasma studies where biotinylated peptides can be easily purified by binding to immobilized avidin beads. FP-Biotin demonstrates good reactivity with human AChE and BChE and is suitable for identifying organophosphorus toxicants reactive proteins.
  • Potent organophosphorus toxicant
  • Suitable for identifying new biomarkers of organophosphorus toxicant exposure
  • Quantifies FAAH, ABHD6, and MAG-lipase activity
  • Useful in plasma studies for easy purification of biotinylated peptides
  • Reacts well with human AChE and BChE
  • Identifies organophosphorus toxicants reactive proteins
Catalog No. 50-003-50282 Supplier Medchemexpress LLC Supplier No. HY13692410MM/1ML
FP-Biotin is a potent organophosphorus toxicant suitable for identifying new biomarkers of organophosphorus toxicant exposure. It quantifies the activity of FAAH, ABHD6, and MAG-lipase. It is used in plasma studies where biotinylated peptides can be easily purified by binding to immobilized avidin beads. FP-Biotin demonstrates good reactivity with human AChE and BChE and is suitable for identifying organophosphorus toxicants reactive proteins.
  • Potent organophosphorus toxicant
  • Suitable for identifying new biomarkers of organophosphorus toxicant exposure
  • Quantifies FAAH, ABHD6, and MAG-lipase activity
  • Useful in plasma studies for easy purification of biotinylated peptides
  • Reacts well with human AChE and BChE
  • Identifies organophosphorus toxicants reactive proteins