Learn More
Medchemexpress LLC FP-biotin | 259270-28-5 | 99.8% | C27H50FN4O5PS | 10MG
SDP
Catalog No. 5000259020
Encompass_Preferred
FP-Biotin is a biotinylated fluorophosphonate probe designed to covalently label serine hydrolases for activity-based profiling. The biotin tag enables enrichment and identification of labeled proteins or peptides from plasma and complex samples using avidin/streptavidin capture. Available as a solid and as a DMSO solution for flexible workflows.
  • covalently labels serine hydrolases for activity profiling
  • enables affinity purification via biotin-avidin interaction
  • suitable for plasma and complex biological samples
  • supplied as solid or ready-to-use solution for workflow flexibility
  • high purity for reproducible results
  • supported by data sheet, COA, and SDS for quality and safety
Catalog No. 50-002-59020 Supplier Medchemexpress LLC Supplier No. HY13692410MG
FP-Biotin is a biotinylated fluorophosphonate probe designed to covalently label serine hydrolases for activity-based profiling. The biotin tag enables enrichment and identification of labeled proteins or peptides from plasma and complex samples using avidin/streptavidin capture. Available as a solid and as a DMSO solution for flexible workflows.
  • covalently labels serine hydrolases for activity profiling
  • enables affinity purification via biotin-avidin interaction
  • suitable for plasma and complex biological samples
  • supplied as solid or ready-to-use solution for workflow flexibility
  • high purity for reproducible results
  • supported by data sheet, COA, and SDS for quality and safety