Learn More
BUCHI FlashPure Empty Solid Loader: Parts
SDP
Catalog No. 05410134 Shop All BUCHI Products
Change view
Click to view available options
Includes:
10 Pc.
20 Pc.
SS male luer lock, PEEK adapter, O-rings
Type:
Adapter Kit
Frit
Size (Metric) Tube Rack:
20 g
80 g
Various
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Type Size (Metric) Tube Rack
05-410-134 SS male luer lock, PEEK adapter, O-rings Adapter Kit Various
05-410-136 20 Pc. Frit 20 g
05-410-129 10 Pc. Frit 80 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-410-134 Supplier BUCHI Supplier No. 145173491
Parts for FlashPure Empty Solid Loaders. FlashPure Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Type Adapter Kit
Includes SS male luer lock, PEEK adapter, O-rings
Size (Metric) Tube Rack Various
For Use With (Equipment) BUCHI and other Luer Lock systems