Learn More
Biotium Fast Plus EvaGreen qPCR Master Mix with Low Rox (5000 rxn)
Catalog No. 501964536
Encompass_Preferred
Fast-Plus EvaGreen Master Mix is a ready-to-use hot-start mix for nucleic acid quantitation and melt analysis of PCR amplicons. It delivers clean PCR product even with the most challenging samples tested. The mix contains environmentally safe EvaGreen dye and fast-activating chemically-modified hotstart enzyme, Cheetah™ Taq. Although the master mix is formulated for qPCR using a fast cycling protocol, it is also compatible with qPCR using a regular cycling protocol. Low ROX reference dye is included in the mix for well-to-well fluorescence normalization in ABI and other instrument platforms. This product has sufficient reagent for 5000 reactions.
Catalog No. 50-196-4536 Supplier Biotium Supplier No. 310142
Fast-Plus EvaGreen Master Mix is a ready-to-use hot-start mix for nucleic acid quantitation and melt analysis of PCR amplicons. It delivers clean PCR product even with the most challenging samples tested. The mix contains environmentally safe EvaGreen dye and fast-activating chemically-modified hotstart enzyme, Cheetah™ Taq. Although the master mix is formulated for qPCR using a fast cycling protocol, it is also compatible with qPCR using a regular cycling protocol. Low ROX reference dye is included in the mix for well-to-well fluorescence normalization in ABI and other instrument platforms. This product has sufficient reagent for 5000 reactions.