Learn More
Biotium Fast Plus EvaGreen qPCR Master Mix (500 rxn)
Catalog No. 501964541
Encompass_Preferred
Fast Plus EvaGreen qPCR master mix is a ready-to-use hot-start mix for nucleic acid quantitation and melt curve analysis of PCR amplicons. Formulated using our environmentally safe EvaGreen dye and fast-activating chemically-modified hotstart enzyme, Cheetah™ Taq. This master mix is offered with ROX reference dye and has sufficient reagent for 500 reactions.
Catalog No. 50-196-4541 Supplier Biotium Supplier No. 310201
Fast Plus EvaGreen qPCR master mix is a ready-to-use hot-start mix for nucleic acid quantitation and melt curve analysis of PCR amplicons. Formulated using our environmentally safe EvaGreen dye and fast-activating chemically-modified hotstart enzyme, Cheetah™ Taq. This master mix is offered with ROX reference dye and has sufficient reagent for 500 reactions.