Learn More
Beckman Coulter Isoton II Diluent 10 Liter
Catalog No. NC2470899 Shop All Beckman Coulter Products
Encompass
Buffered saline with preservatives isotonic diluent and sheath fluid for hematology applications on analyzers, maintains cell volume and size for accurate hemocytometry and cell counting.
Catalog No. NC2470899 Supplier Beckman Coulter Supplier No. C96980
Buffered saline with preservatives isotonic diluent and sheath fluid for hematology applications on analyzers, maintains cell volume and size for accurate hemocytometry and cell counting.