Learn More
Biotium Fast Probe Master Mix with Rox (500 rxn)
Catalog No. 501964540
Encompass_Preferred
Fast Probe Master Mix is formulated for conducting real-time PCR using an oligo-based probe, such as a TaqMan probe, a MGB probe or an AllGlo™ probe. The master mix contains buffer, dNTPs, MgCl2 and our superior hotstart enzyme Cheetah™ Taq. Cheetah™ Taq is a chemically modified hotstart Taq which can be fully activated in 2 minutes, thus permitting fast PCR. This master mix is offered with ROX reference dye and has sufficient reagent for 500 reactions.
Catalog No. 50-196-4540 Supplier Biotium Supplier No. 310161
Fast Probe Master Mix is formulated for conducting real-time PCR using an oligo-based probe, such as a TaqMan probe, a MGB probe or an AllGlo™ probe. The master mix contains buffer, dNTPs, MgCl2 and our superior hotstart enzyme Cheetah™ Taq. Cheetah™ Taq is a chemically modified hotstart Taq which can be fully activated in 2 minutes, thus permitting fast PCR. This master mix is offered with ROX reference dye and has sufficient reagent for 500 reactions.