Learn More
Agilent Technologies FP SAHR 96 800U PE25-GF.7 25CS
Catalog No. 50995943
Encompass
Seahorse polypropylene 96-well filter microplate with 25um polyethylene frit plus 0.7um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-943 Supplier Agilent Technologies Supplier No. 201019100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 25um polyethylene frit plus 0.7um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100