Learn More
Agilent Technologies Microplate, 384-Well, Polypropylene, Long Drip, 140 μL, 10/Pk.
Catalog No. 50995956
Encompass
This 384-well microplate is manufactured from polypropylene and designed for various laboratory applications. It features a long drip design and provides a capacity of 140 μL per well.
  • Constructed from durable polypropylene
  • Features a 384-well format for high-throughput screening
  • Equipped with long drip design for optimized liquid handling
  • Contains no filter
  • Provides 140 μL volume per well
Catalog No. 50-995-956 Supplier Agilent Technologies Supplier No. 201031100
This 384-well microplate is manufactured from polypropylene and designed for various laboratory applications. It features a long drip design and provides a capacity of 140 μL per well.
  • Constructed from durable polypropylene
  • Features a 384-well format for high-throughput screening
  • Equipped with long drip design for optimized liquid handling
  • Contains no filter
  • Provides 140 μL volume per well