Learn More
Agilent Technologies FP SEAHORSE 48 5ML PE 25 25/CS
Supplier:  Agilent Technologies 200923100
Encompass
Seahorse polypropylene 48-well filter microplate with 25um polyethylene frit, 5ml/well, long drip, case of 25, recommended receiver plate 201238-100
Catalog No. 50-995-896