Learn More
Agilent Technologies FP SEAHORSE 96 2ML PE 25 25/CS
Supplier:  Agilent Technologies 200919100
Encompass
Seahorse polypropylene 96-well filter microplate with 25um polyethylene frit, 2ml/well, long drip, case of 25, recommended receiver plate 201240-100
Catalog No. 50-995-895
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.