Learn More
Agilent Technologies FP SEAHORSE 96 400UL 30K 25/CS
Catalog No. 50995941
Encompass
Seahorse polypropylene 96-well filter microplate with 30KDa polyethersulfone membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-941 Supplier Agilent Technologies Supplier No. 201015100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 30KDa polyethersulfone membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100